Learn More
BMP-1/PCP Rat anti-Human, Alexa Fluor™ 350, Clone: MM0112-11B23, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212055AF350
Catalog No. NP2120A30
Add to cart
Description
Description
BMP-1/PCP Monoclonal antibody specifically detects BMP-1/PCP in Human samples. It is validated for Western Blot.
Specifications
Specifications
BMP-1/PCP
MM0112-11B23
Western Blot
BMP-1, bone morphogenetic protein 1, EC 3.4.24, EC 3.4.24.19, FLJ44432, Mammalian tolloid protein, mTld, PCOLC, PCP, PCP2, procollagen C-endopeptidase, Procollagen C-proteinase, TLD
Rat
IgG2
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
BMP1
Human BMP1
0.1 mL
Primary
649