Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMP-1/PCP Rat anti-Human, Alexa Fluor™ 750, Clone: MM0112-11B23, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP212055AF750

Catalog No. NP2120A70


Add to cart

Description

Description

BMP-1/PCP Monoclonal antibody specifically detects BMP-1/PCP in Human samples. It is validated for Western Blot.
Specifications

Specifications

BMP-1/PCP
MM0112-11B23
Western Blot
Conjugated, Purified
BMP1
Human BMP1
Protein G purified
Cytokine Research
Primary
649.0
Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
BMP-1, bone morphogenetic protein 1, EC 3.4.24, EC 3.4.24.19, FLJ44432, Mammalian tolloid protein, mTld, PCOLC, PCP, PCP2, procollagen C-endopeptidase, Procollagen C-proteinase, TLD
Rat
IgG2
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit