Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BMP-10 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$176.85 - $666.00
Specifications
Antigen | BMP-10 |
---|---|
Concentration | 5.0 mg/mL |
Dilution | ELISA 1:100-1:2000 |
Applications | ELISA |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10062392
|
Novus Biologicals
NB10062392 |
0.5 mL |
Each for $666.00
|
|
NB017467
|
Novus Biologicals
NB100623920.1ML |
0.1 mL |
Each for $176.85
|
|
Description
BMP-10 Polyclonal specifically detects BMP-10 in Human samples. It is validated for ELISA.Specifications
BMP-10 | |
ELISA 1:100-1:2000 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
O95393 | |
27302 | |
Synthetic peptide corresponding to the amino acids 84 - 94 of human BMP-10 (402-412 of the unprocessed precursor) conjugated to Keyhole Limpet Haemocyanin. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
5.0 mg/mL | |
ELISA | |
Unconjugated | |
Rabbit | |
Cell Cycle and Replication | |
PBS with 0.09% Sodium Azide | |
BMP-10, bone morphogenetic protein 10, MGC126783 | |
BMP10 | |
IgG | |
Protein G purified | |
BMP-10 |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title