Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BMP-15/GDF-9B Mouse anti-Human, Clone: 28A, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP261935
Description
BMP-15/GDF-9B Monoclonal antibody specifically detects BMP-15/GDF-9B in Human samples. It is validated for Western Blot, ELISA.Specifications
BMP-15/GDF-9B | |
28A | |
Unconjugated | |
PBS with No Preservative | |
BMP-15, bone morphogenetic protein 15, GDF-9B, GDF9BODG2, Growth/differentiation factor 9B, POF4 | |
Mouse | |
IgG1 | |
Protein A purified | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
Monoclonal | |
Human |
ELISA, Western Blot | |
1 mg/mL | |
Western Blot, ELISA | |
Purified | |
BMP15 | |
Recognizes sequence SAEVTASSSKHSGPENNQC on the C-terminus of human GDF9B (BMP15) | |
55 kDa | |
0.1 mg | |
Primary | |
9210 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title