Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMP-15/GDF-9B Mouse anti-Human, Clone: BMP15/4263, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen BMP-15/GDF-9B
Clone BMP15/4263
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, ELISA
Applications Western Blot, ELISA
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122577
View Documents
Novus Biologicals
NBP30755210UG
100 μg
Each for $352.89
Add to cart
 
NB122578
View Documents
Novus Biologicals
NBP30755220UG
20 μg
Each for $138.37
Add to cart
 
Description

Description

BMP-15/GDF-9B Monoclonal antibody specifically detects BMP-15/GDF-9B in Human samples. It is validated for Western Blot, ELISA
Specifications

Specifications

BMP-15/GDF-9B
0.2 mg/ml
Western Blot, ELISA
Purified
RUO
10 mM PBS with 0.05% BSA
9210
IgG1
Primary
Store at 4°C.
BMP15/4263
Western Blot 1-2 ug/ml, ELISA
Unconjugated
Mouse
Human
BMP-15, bone morphogenetic protein 15, GDF-9B, Growth/differentiation factor 9B, ODG2, POF4
Recognizes sequence SAEVTASSSKHSGPENNQC on the C-terminus of human BMP-15/GDF-9B
Monoclonal
Protein A or G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit