Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BMP-15/GDF-9B Mouse anti-Human, DyLight 405, Clone: 28A, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP261935V
Description
BMP-15/GDF-9B Monoclonal antibody specifically detects BMP-15/GDF-9B in Human samples. It is validated for Western Blot, ELISA.Specifications
BMP-15/GDF-9B | |
28A | |
Western Blot, ELISA | |
50mM Sodium Borate with 0.05% Sodium Azide | |
9210 | |
Mouse | |
IgG1 | |
Primary | |
0.1 ml | |
Human |
Western Blot,ELISA | |
DyLight 405 | |
Purified | |
BMP-15, bone morphogenetic protein 15, GDF-9B, GDF9BODG2, Growth/differentiation factor 9B, POF4 | |
BMP15 | |
Recognizes sequence SAEVTASSSKHSGPENNQC on the C-terminus of human GDF9B (BMP15) | |
Monoclonal | |
Protein A purified | |
Store at 4C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title