Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMP-15/GDF-9B Mouse anti-Human, DyLight 550, Clone: 28A, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP261935R

Catalog No. NBP261935R


Add to cart

Description

Description

BMP-15/GDF-9B Monoclonal antibody specifically detects BMP-15/GDF-9B in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

BMP-15/GDF-9B
28A
Western Blot, ELISA
Purified
BMP15
Recognizes sequence SAEVTASSSKHSGPENNQC on the C-terminus of human GDF9B (BMP15)
Protein A purified
Store at 4C in the dark.
Monoclonal
Human
ELISA, Western Blot
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
BMP-15, bone morphogenetic protein 15, GDF-9B, GDF9BODG2, Growth/differentiation factor 9B, POF4
Mouse
IgG1
0.1 ml
Primary
9210
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit