Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ BMP-2 Antibody Pair [HRP]

Mouse Monoclonal Antibody Pair

Manufacturer:  Novus Biologicals™ NBP2795725PLATES

 View more versions of this product

Catalog No. NB010184


Add to cart

Description

Description

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human BMP-2.
Specifications

Specifications

Sandwich ELISA
Antibody Pairs
31.25 pg/mL
650
RUO
BMP-2
Biologically Active Proteins, Cytokine Research, Immunology, Neuronal Cell Markers, Stem Cell Markers, Stem Cell Signaling Pathway
Human
BMP2
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 1 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 2000 pg/mL is recommended.
HRP
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human BMP-2.
Sandwich ELISA
  • Standard: Each vial contains 35 ng of recombinant Human BMP-2.
  • Capture Antibody: 1 mg/mL of mouse anti-Human BMP-2 monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.2 mg/mL of mouse anti-Human BMP-2 monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Storage is content dependent.
    Mouse
    5 Plates
    31.25-2000 pg/mL
    BMP-2, BMP-2A, BMP2ABone morphogenetic protein 2A, bone morphogenetic protein 2
    Conjugated
    SDS
    Documents
    Product Certifications
    Promotions

    Promotions

    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit