Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BMP-2 Antibody (15B3), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242651AF488
Description
BMP-2 Monoclonal specifically detects BMP-2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
BMP-2 | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
BMP2 | |
Recombinant human BMP2 (283-396aa) purified from E. coli | |
100 μL | |
Biologically Active Proteins, Cytokine Research, Embryonic Stem Cell Markers, Immunology, Neuronal Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers, Stem Cell Signaling Pathway | |
650 | |
Store at 4C in the dark. | |
IgG2b κ |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
15B3 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
BMP-2, BMP-2A, BMP2ABone morphogenetic protein 2A, bone morphogenetic protein 2 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction