Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BMP-7 Mouse anti-Human, DyLight 755, Clone: 4E7, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP104259IR
Description
BMP-7 Monoclonal specifically detects BMP-7 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
BMP-7 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
BMP7 | |
Recombinant human BMP7 (293-431 aa) purified from E. coli | |
0.1 mL | |
Biologically Active Proteins, Cytokine Research | |
655 | |
Human | |
Purified |
Western Blot, ELISA | |
40000000 | |
Western Blot, ELISA | |
BMP-7, bone morphogenetic protein 7, Eptotermin alfa, OP-1OP1, Osteogenic protein 1 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Anti-human BMP7 mAb, clone 4E7, is derived from hybridization of mouse SP2/O myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human BMP7 protein. | |
Store at 4C in the dark. | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title