Learn More
Brady™ BMP™71 Series WorkHorse™ Permanent Polyester Labels - White
Permanent Polyester (B-423) is ideal for electronic PCB and component identification, bar code labels, rating plates and solar panel identification.
$37.05 - $48.00
Products 2
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
19-000-145 View Documents Brady™
M7119423
1 in. H x 1 in. W Each for $48.00
Add to cart
19-000-146 View Documents Brady™
M7120423
1 in. H x 2 in. W Each for $37.05
Add to cart
Description
Description
  • Glossy white finish with permanent acrylic is thermal transfer printable
  • RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC
  • UL approved
  • CSA accepted
  • Meets requirements of a halogen-free material per DIN VDE 0472 part 815
  • Service temperature of -94 to 230° F (-70 to 110° C)