Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Brady™ BMP™71 TLS 2200™ Series Durasleeve™ Rigid Polypropylene Wire Marker Inserts - White
DuraSleeve™ Polypropylene Inserts (B-390) are printable, rigid inserts designed to be printed and affixed to a wire using extruded, clear PVC Wire Marking Carriers.
$761.00 - $856.00
Products 2
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
19-109-038 View Documents Brady™
BPTDS15390
0.163 in. H x 0.59 in. W Each for $761.00
N/A
19-109-039 View Documents Brady™
BPTDS30390
0.163 in. H x 1.181 in. W Each for $856.00
N/A
Description
Description
  • Inserts are supplied roll form formatted for thermal transfer printing on the TLS 2200™ Thermal Labeling System and benchtop printers
  • B-390 material meets the requirements of a halogen-free material per DIN VDE 0472 part 815
  • RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC
  • Service temperature of -40°F to 212°F (-40°C to 100°C)