Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMP-8b Mouse anti-Human, Alexa Fluor™ 532, Clone: 13000000, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP242693AF532

Catalog No. NP242693A32


Add to cart

Description

Description

BMP-8b Monoclonal antibody specifically detects BMP-8b in Human samples. It is validated for Western Blot,ELISA,Immunocytochemistry,Immunofluorescence.
Specifications

Specifications

BMP-8b
13000000
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
BMP-8, bone morphogenetic protein 8b, MGC131757
Mouse
IgG2a Kappa
Store at 4°C in the dark.
Monoclonal
Human
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
BMP8B
Recombinant human BMP8b (264-402aa) purified from E. coli
0.1 mL
Primary
656
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit