Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BMP-8b Antibody (13E6), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242693AF750
Description
BMP-8b Monoclonal specifically detects BMP-8b in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
BMP-8b | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
BMP8B | |
Recombinant human BMP8b (264-402aa) purified from E. coli | |
Primary | |
Human | |
IgG2a κ |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
13000000 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
BMP-8, bone morphogenetic protein 8b, MGC131757 | |
Mouse | |
100 μL | |
656 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction