Learn More
BMP-9 Mouse anti-Human, Alexa Fluor™ 532, Clone: 16S11, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP222034AF532
Catalog No. NP222034A32
Add to cart
Description
Description
BMP-9 Monoclonal antibody specifically detects BMP-9 in Human samples. It is validated for Western Blot,ELISA.
Specifications
Specifications
BMP-9
16S11
Western Blot, ELISA
BMP9BMP-9Bone morphogenetic protein 9, GDF-2, growth differentiation factor 2, growth/differentiation factor 2
Mouse
IgG2
Store at 4°C in the dark.
Monoclonal
Human
ELISA, Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
GDF2
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with purified recombinant BMP-9. Mouse anti human BMP-9 Q9UK05.
0.1 mL
Primary
2658