Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMP-9 Mouse anti-Human, Alexa Fluor™ 594, Clone: 16S11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP222034AF594

Catalog No. NP222034A94


Add to cart

Description

Description

BMP-9 Monoclonal antibody specifically detects BMP-9 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

BMP-9
16S11
Western Blot, ELISA
Conjugated, Purified
GDF2
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with purified recombinant BMP-9. Mouse anti human BMP-9 Q9UK05.
Protein G purified
Cytokine Research
Primary
2658.0
ELISA, Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
BMP9BMP-9Bone morphogenetic protein 9, GDF-2, growth differentiation factor 2, growth/differentiation factor 2
Mouse
IgG2
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit