Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BMP-9 Mouse anti-Human, HRP, Clone: 16S11, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222034H
Description
BMP-9 Monoclonal specifically detects BMP-9 in Human samples. It is validated for Western Blot, ELISA.Specifications
BMP-9 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
GDF2 | |
Human recombinant BMP-9 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, ELISA | |
16S11 | |
Western Blot, ELISA | |
BMP9BMP-9Bone morphogenetic protein 9, GDF-2, growth differentiation factor 2, growth/differentiation factor 2 | |
Mouse | |
Protein G purified | |
Cytokine Research | |
2658 | |
Store at 4C in the dark. | |
IgG2 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title