Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BMP1 Mouse anti-Human, Clone: OTI3E9, liquid, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene TA505813
Description
BMP1 encodes a protein that is capable of inducing formation of cartilage in vivo. Although other bone morphogenetic proteins are members of the TGF-beta superfamily, this gene encodes a protein that is not closely related to other known growth factors. This gene is expressed as alternatively spliced variants that share an N-terminal protease domain but differ in their C-terminal region.Specifications
BMP1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P13497 | |
OI13, PCOLC, PCP, PCP2, TLD | |
Mouse | |
IgG1 | |
100 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
649 |
Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI3E9 | |
Unconjugated | |
BMP1 | |
Liquid | |
BMP1 | |
Human recombit protein fragment corresponding to amino acids 225-617 of human BMP1 produced in E.coli. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title