Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BMPR-IA/ALK-3 Mouse anti-Human, Alexa Fluor™ 594, Clone: MM0053-8L7, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212029AF594
Description
BMPR-IA/ALK-3 Monoclonal specifically detects BMPR-IA/ALK-3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
BMPR-IA/ALK-3 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
BMPR1A | |
Human recombinant BMPR-IA/ALK-3 (extracellular domain) | |
0.1 mL | |
Primary | |
In Western blots, this antibody shows good cross-reactivity with mouse BMPR-IA/ALK-3, but no cross-reactivity with recombinant human BMPR-IB or rhBMPR-II. | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
MM0053-8L7 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
ACVRLK310q23del, ALK-3, ALK3EC 2.7.11.30, BMP type-1A receptor, BMPR-1A, bone morphogenetic protein receptor type-1A, bone morphogenetic protein receptor, type IA, CD292, CD292 antigen, EC 2.7.11, Serine/threonine-protein kinase receptor R5, SKR5, type II-like kinase 3 | |
Mouse | |
Protein G purified | |
Mesenchymal Stem Cell Markers, Protein Kinase, Stem Cell Markers | |
657.0 | |
Human | |
IgG2 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title