Learn More
BMPR-IB/ALK-6 Mouse anti-Human, Alexa Fluor 488, Clone: MM0055-3E12, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP212031AF488
Catalog No. NB212031488
Add to cart
Description
Description
BMPR-IB/ALK-6 Monoclonal specifically detects BMPR-IB/ALK-6 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
BMPR-IB/ALK-6
MM0055-3E12
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
BMPR1B
Human BMPR1B
Protein G purified
Mesenchymal Stem Cell Markers, Protein Kinase, Stem Cell Markers
Primary
658
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
activin receptor-like kinase 6, ALK-6, ALK6CDw293, BMP type-1B receptor, BMPR-1B, bone morphogenetic protein receptor type-1B, bone morphogenetic protein receptor, type IB, CDw293 antigen, EC 2.7.11, EC 2.7.11.30, serine/threonine receptor kinase
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human