Learn More
BMPR-II Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Manufacturer:  R&D Systems AF811SP
Catalog No. AF811SP
Add to cart
This item is not returnable. View return policy
Description
Description
BMPR-II Polyclonal antibody AF811SP specifically detects BMPR-II in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Western Blotting
Specifications
Specifications
BMPR-II
LYOPH
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, CyTOF-ready
Q13873
BMP type II receptor, BMP type-2 receptor, BMPR2, BMPR-2, BMPR3, BMPRII, BMPR-IIprimary pulmonary hypertension 1, Bone morphogenetic protein receptor type II, bone morphogenetic protein receptor type-2, bone morphogenetic protein receptor, type II (serine/threonine kinase), BRK-3, EC 2.7.11, EC 2.7.11.30, FLJ41585, FLJ76945, PPH1BMR2, T-ALK, type II activin receptor-like kinase, type II receptor for bone morphogenetic protein-4
Goat
IgG
25 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human BMPR-II in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) BMPR-IA, recombinant mouse (rm) BMPR-IA, rhBMPR-IB, and rmBMPR-IB is observed.
Human
CyTOF, Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
BMPR2
Mouse myeloma cell line NS0-derived recombinant human BMPR-II Ala26-Ile151 Accession # Q13873
Immunogen affinity purified
RUO
Primary
659
Reconstitute at 0.2 mg/mL in sterile PBS.