Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BMS 182874 hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent, selective and competitive non-peptide endothelin ETA receptor antagonist (Ki = 48 nM). Displays > 1000-fold selectivity over ETB receptors. Inhibits ET-1-induced pressor response following oral or intravenous administration in vivo. Inhibits ET-1-induced longitudinal muscle contraction in the mouse colon in vitro.
Specifications
Quantity | 50 mg |
Formula Weight | 381.88 |
Percent Purity | >98% |
Chemical Name or Material | BMS 182874 hydrochloride |
Safety and Handling
Recommended Storage | Store at Room Temperature |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction