Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMS 182874 hydrochloride, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg
1215703-04-0
C17H20ClN3O3S
381.875
UZZFRNZGYDOBAO-UHFFFAOYSA-N
bms 182874 hydrochloride,5-dimethylamino-n-3,4-dimethyl-5-isoxazolyl-1-naphthalenesulfonamide hydrochloride
56972163
5-(dimethylamino)-N-(3,4-dimethyl-1,2-oxazol-5-yl)naphthalene-1-sulfonamide;hydrochloride
CC1=C(ON=C1C)NS(=O)(=O)C2=CC=CC3=C2C=CC=C3N(C)C.Cl
1215703-04-0
C17H20ClN3O3S
381.875
UZZFRNZGYDOBAO-UHFFFAOYSA-N
bms 182874 hydrochloride,5-dimethylamino-n-3,4-dimethyl-5-isoxazolyl-1-naphthalenesulfonamide hydrochloride
56972163
5-(dimethylamino)-N-(3,4-dimethyl-1,2-oxazol-5-yl)naphthalene-1-sulfonamide;hydrochloride
CC1=C(ON=C1C)NS(=O)(=O)C2=CC=CC3=C2C=CC=C3N(C)C.Cl

Highly selective, orally active non-peptide ETA antagonist

Potent, selective and competitive non-peptide endothelin ETA receptor antagonist (Ki = 48 nM). Displays > 1000-fold selectivity over ETB receptors. Inhibits ET-1-induced pressor response following oral or intravenous administration in vivo. Inhibits ET-1-induced longitudinal muscle contraction in the mouse colon in vitro.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
Formula Weight 381.88
Percent Purity >98%
Chemical Name or Material BMS 182874 hydrochloride

Recommended Storage : Store at Room Temperature