Learn More
Tocris Bioscience™ BMS 509744
Potent and selective ITK inhibitor
Supplier:  Tocris Bioscience™ 5009/10
Catalog No. 50-091-0R
Add to Cart
This item is not returnable. View return policy
Description
Description
BMS 509744 is a potent and selective interleukin-2 inducible T cell kinase (ITK) inhibitor (IC50 = 19 nM). Displays 200-fold selectivity over Tec family kinases and 55-fold selectivity over other kinases tested. Reduces HIV infection of primary CD4+ T cells and attenuates the establishment of HIV infection in vitro. Reduces T cell proliferation and IL-2 production in vitro. Reduces lung inflammation in a mouse model of ovalbumin-induced allergy/asthma.
Specifications
Specifications
97%
439575-02-7
10 mg
M.W. (Observed): 623.83
N-[5-[[5-[(4-Acetyl-1-piperazinyl)carbonyl]-4-methoxy-2-methylphenyl]thio]-2-thiazolyl]-4-[[(1,2,2-trimethylpropyl)amino]methyl]benzamide
-20°C
BMS 509744
BMS 509744