Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BMS 536924, Tocris Bioscience™
Click to view available options
Quantity:
10mg
50mg
Description
Specifications
Quantity | 50mg |
Formula Weight | 479.96 |
Percent Purity | >98% |
Chemical Name or Material | BMS 536924 |
Safety and Handling
Recommended Storage : Store at -20°C