Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BMS 566419

Inosine monophosphatase dehydrogenase (IMPDH) inhibitor

$203.50 - $843.50

Specifications

Chemical Name or Material N-[1-[6-(4-Ethyl-1-piperazinyl)-3-pyridinyl]-1-methylethyl]-2-fluoro-9,10-dihydro-9-oxo-3-acridinecarboxamide
Molecular Formula C28H30FN5O2
CAS 566161-24-8
Product Type Target: Dehydrogenase Inhibitors
Formula Weight 487.57
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
549250 View Documents Tocris Bioscience™
5492/50
50mg Each for $843.50
Add to cart
 
549210 View Documents Tocris Bioscience™
5492/10
10mg Each for $203.50
Add to cart
 
Description

Description

Potent inosine monophosphate dehydrogenase (IMPDH) inhibitor (IC50 = 17 nM). Inhibits proliferation of human T-lymphoblasts and PBMCs in vitro. Reduces paw swelling in a rat adjuvant arthritis model. Orally available.
Specifications

Specifications

N-[1-[6-(4-Ethyl-1-piperazinyl)-3-pyridinyl]-1-methylethyl]-2-fluoro-9,10-dihydro-9-oxo-3-acridinecarboxamide
566161-24-8
487.57
C28H30FN5O2
Target: Dehydrogenase Inhibitors
Safety and Handling

Safety and Handling

Recommended Storage : Store at 4°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit