Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMY 14802 hydrochloride, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
105565-55-7
C18H23ClF2N4O
384.856
NIBVEFRJDFVQLM-UHFFFAOYSA-N
bmy 14802 hydrochloride,bmy-14802 hydrochloride,bmy 14802 hcl,hcl,1-4-fluorophenyl-4-4-5-fluoropyrimidin-2-yl piperazin-1-yl butan-1-ol hydrochloride,1-piperazinebutanol, alpha-4-fluorophenyl-4-5-fluoro-2-pyrimidinyl-, monohydrochloride,1-4-fluorophenyl-4-4-5-fluoro-2-pyrimidinyl-1-piperazinyl-1-butanol monohydrochloride,1-4-fluorophenyl-4-4-5-fluoropyrimidin-2-yl piperazin-1-yl butan-1-ol hydrochloride;1-4-fluorophenyl-4-4-5-fluoropyrimidin-2-yl piperazin-1-yl butan-1-ol hydrochloride
3086514
1-(4-fluorophenyl)-4-[4-(5-fluoropyrimidin-2-yl)piperazin-1-yl]butan-1-ol;hydrochloride
C1CN(CCN1CCCC(C2=CC=C(C=C2)F)O)C3=NC=C(C=N3)F.Cl
105565-55-7
C18H23ClF2N4O
384.856
NIBVEFRJDFVQLM-UHFFFAOYSA-N
bmy 14802 hydrochloride,bmy-14802 hydrochloride,bmy 14802 hcl,hcl,1-4-fluorophenyl-4-4-5-fluoropyrimidin-2-yl piperazin-1-yl butan-1-ol hydrochloride,1-piperazinebutanol, alpha-4-fluorophenyl-4-5-fluoro-2-pyrimidinyl-, monohydrochloride,1-4-fluorophenyl-4-4-5-fluoro-2-pyrimidinyl-1-piperazinyl-1-butanol monohydrochloride,1-4-fluorophenyl-4-4-5-fluoropyrimidin-2-yl piperazin-1-yl butan-1-ol hydrochloride;1-4-fluorophenyl-4-4-5-fluoropyrimidin-2-yl piperazin-1-yl butan-1-ol hydrochloride
3086514
1-(4-fluorophenyl)-4-[4-(5-fluoropyrimidin-2-yl)piperazin-1-yl]butan-1-ol;hydrochloride
C1CN(CCN1CCCC(C2=CC=C(C=C2)F)O)C3=NC=C(C=N3)F.Cl

Sigma antagonist. Antipsychotic agent

Potent sigma receptor antagonist (IC50 = 112 nM) with modest to weak affinity for 5-HT1A and α1 receptors. Antipsychotic following oral administration and acts via indirect modulation of central dopaminergic systems.
Quantity 50mg
Formula Weight 384.86
Percent Purity >98%
Chemical Name or Material BMY 14802 hydrochloride

Recommended Storage : Desiccate at Room Temperature