Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMY 14802 hydrochloride, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg
105565-55-7
C18H23ClF2N4O
384.856
NIBVEFRJDFVQLM-UHFFFAOYSA-N
bmy 14802 hydrochloride,bmy-14802 hydrochloride,bmy 14802 hcl,hcl,1-4-fluorophenyl-4-4-5-fluoropyrimidin-2-yl piperazin-1-yl butan-1-ol hydrochloride,1-piperazinebutanol, alpha-4-fluorophenyl-4-5-fluoro-2-pyrimidinyl-, monohydrochloride,1-4-fluorophenyl-4-4-5-fluoro-2-pyrimidinyl-1-piperazinyl-1-butanol monohydrochloride,1-4-fluorophenyl-4-4-5-fluoropyrimidin-2-yl piperazin-1-yl butan-1-ol hydrochloride;1-4-fluorophenyl-4-4-5-fluoropyrimidin-2-yl piperazin-1-yl butan-1-ol hydrochloride
3086514
1-(4-fluorophenyl)-4-[4-(5-fluoropyrimidin-2-yl)piperazin-1-yl]butan-1-ol;hydrochloride
C1CN(CCN1CCCC(C2=CC=C(C=C2)F)O)C3=NC=C(C=N3)F.Cl
105565-55-7
C18H23ClF2N4O
384.856
NIBVEFRJDFVQLM-UHFFFAOYSA-N
bmy 14802 hydrochloride,bmy-14802 hydrochloride,bmy 14802 hcl,hcl,1-4-fluorophenyl-4-4-5-fluoropyrimidin-2-yl piperazin-1-yl butan-1-ol hydrochloride,1-piperazinebutanol, alpha-4-fluorophenyl-4-5-fluoro-2-pyrimidinyl-, monohydrochloride,1-4-fluorophenyl-4-4-5-fluoro-2-pyrimidinyl-1-piperazinyl-1-butanol monohydrochloride,1-4-fluorophenyl-4-4-5-fluoropyrimidin-2-yl piperazin-1-yl butan-1-ol hydrochloride;1-4-fluorophenyl-4-4-5-fluoropyrimidin-2-yl piperazin-1-yl butan-1-ol hydrochloride
3086514
1-(4-fluorophenyl)-4-[4-(5-fluoropyrimidin-2-yl)piperazin-1-yl]butan-1-ol;hydrochloride
C1CN(CCN1CCCC(C2=CC=C(C=C2)F)O)C3=NC=C(C=N3)F.Cl

Sigma antagonist. Antipsychotic agent

Potent sigma receptor antagonist (IC50 = 112 nM) with modest to weak affinity for 5-HT1A and α1 receptors. Antipsychotic following oral administration and acts via indirect modulation of central dopaminergic systems.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
Formula Weight 384.86
Percent Purity >98%
Chemical Name or Material BMY 14802 hydrochloride

Recommended Storage : Desiccate at Room Temperature