Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMY 45778, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
152575-66-1
C26H18N2O5
438.439
DSRSEEYZGWTODH-UHFFFAOYSA-N
3-4,5-diphenyl 2,4'-bioxazol-5'-yl phenoxy acetic acid,3-4,5-diphenyl-2,4'-bi-1,3-oxazol-5'-yl phenoxy acetic acid,tocris-1442,d0u1sf,3-4-4,5-diphenyl-2-oxazolyl-5-oxazolyl phenoxy acetic acid,acetic acid, 3-4,5-diphenyl 2,4'-bioxazol-5'-yl phenoxy,2-3-4-4,5-diphenyl-1,3-oxazol-2-yl-1,3-oxazol-5-yl phenoxy acetic acid,2-3-4-4,5-di phenyl-1,3-oxazol-2-yl-1,3-oxazol-5-yl phenoxy acetic acid
127861
CHEBI:51544
2-[3-[4-(4,5-diphenyl-1,3-oxazol-2-yl)-1,3-oxazol-5-yl]phenoxy]acetic acid
C1=CC=C(C=C1)C2=C(OC(=N2)C3=C(OC=N3)C4=CC(=CC=C4)OCC(=O)O)C5=CC=CC=C5
This item is not returnable. View return policy
152575-66-1
C26H18N2O5
438.439
DSRSEEYZGWTODH-UHFFFAOYSA-N
3-4,5-diphenyl 2,4'-bioxazol-5'-yl phenoxy acetic acid,3-4,5-diphenyl-2,4'-bi-1,3-oxazol-5'-yl phenoxy acetic acid,tocris-1442,d0u1sf,3-4-4,5-diphenyl-2-oxazolyl-5-oxazolyl phenoxy acetic acid,acetic acid, 3-4,5-diphenyl 2,4'-bioxazol-5'-yl phenoxy,2-3-4-4,5-diphenyl-1,3-oxazol-2-yl-1,3-oxazol-5-yl phenoxy acetic acid,2-3-4-4,5-di phenyl-1,3-oxazol-2-yl-1,3-oxazol-5-yl phenoxy acetic acid
127861
CHEBI:51544
2-[3-[4-(4,5-diphenyl-1,3-oxazol-2-yl)-1,3-oxazol-5-yl]phenoxy]acetic acid
C1=CC=C(C=C1)C2=C(OC(=N2)C3=C(OC=N3)C4=CC(=CC=C4)OCC(=O)O)C5=CC=CC=C5

Non-prostanoid prostacyclin IP receptor partial agonist

Non-prostanoid prostacyclin mimetic that acts as a partial agonist at IP1 prostacyclin receptors. Potently inhibits platelet aggregation in vitro (IC50 = 27-35 nM).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
Formula Weight 438.44
Percent Purity >99%
Chemical Name or Material BMY 45778

Recommended Storage : Store at +4°C