Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BMY 45778, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Non-prostanoid prostacyclin mimetic that acts as a partial agonist at IP1 prostacyclin receptors. Potently inhibits platelet aggregation in vitro (IC50 = 27-35 nM).
Specifications
Quantity | 50 mg |
Formula Weight | 438.44 |
Percent Purity | >99% |
Chemical Name or Material | BMY 45778 |
Safety and Handling
Recommended Storage | Store at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction