Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMY 7378 dihydrochloride, Tocris Bioscience™

Catalog Number 100650
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
21102-95-4
C22H33Cl2N3O3
458.424
NIBOMXUDFLRHRV-UHFFFAOYSA-N
bmy 7378 dihydrochloride, 8-2-4-2-methoxyphenyl piperazin-1-yl ethyl-8-azaspiro 4.5 decane-7,9-dione dihydrochloride, unii-kc07kv8t5o, kc07kv8t5o, 1,1-cyclopentanediacetimide, n-2-4-o-methoxyphenyl-1-piperazinyl ethyl-, dihydrochloride, azaspiro 4.5 decane-7,9-dione dihydrochloride, n-2-4-o-methoxyphenyl-1-piperazinyl ethyl-1,1-cyclopentanediacetimide dihydrochloride, bmy7378 dihydrochloride
210172
8-[2-[4-(2-methoxyphenyl)piperazin-1-yl]ethyl]-8-azaspiro[4.5]decane-7,9-dione;dihydrochloride
COC1=CC=CC=C1N2CCN(CC2)CCN3C(=O)CC4(CCCC4)CC3=O.Cl.Cl
This item is not returnable. View return policy
21102-95-4
C22H33Cl2N3O3
458.424
NIBOMXUDFLRHRV-UHFFFAOYSA-N
bmy 7378 dihydrochloride, 8-2-4-2-methoxyphenyl piperazin-1-yl ethyl-8-azaspiro 4.5 decane-7,9-dione dihydrochloride, unii-kc07kv8t5o, kc07kv8t5o, 1,1-cyclopentanediacetimide, n-2-4-o-methoxyphenyl-1-piperazinyl ethyl-, dihydrochloride, azaspiro 4.5 decane-7,9-dione dihydrochloride, n-2-4-o-methoxyphenyl-1-piperazinyl ethyl-1,1-cyclopentanediacetimide dihydrochloride, bmy7378 dihydrochloride
210172
8-[2-[4-(2-methoxyphenyl)piperazin-1-yl]ethyl]-8-azaspiro[4.5]decane-7,9-dione;dihydrochloride
COC1=CC=CC=C1N2CCN(CC2)CCN3C(=O)CC4(CCCC4)CC3=O.Cl.Cl

Selective α1D antagonist

5-HT1A partial agonist and high affinity α1D adrenoceptor antagonist (Ki values are 2, 800 and 600 nM at cloned rat α1D, rat α1A and hamster α1B receptors, respectively).
Quantity 50 mg
Formula Weight 458.42
Percent Purity >98%
Chemical Name or Material BMY 7378 dihydrochloride
Recommended Storage Store at Room Temperature
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit