Learn More
BNC2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305487A

Description
The recommended shelf life for this product is 1 year from date of receipt.
Probable transcription factor specific for skin keratinocytes. May play a role in the differentiation of spermatozoa and oocytes.Specifications
BNC2 | |
Polyclonal | |
Unconjugated | |
BNC2 | |
Zinc finger protein basonuclin-2, BNC2 (HGNC:30988) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54796 | |
4° C | |
Liquid |
Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q6ZN30 | |
BNC2 | |
Between 350 and 400 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.