Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BNC2 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP169078
Description
BNC2 Polyclonal specifically detects BNC2 in Mouse samples. It is validated for Western Blot.Specifications
BNC2 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
basonuclin 2, BSN2, DKFZp686A01127, FLJ20043, FLJ34928, zinc finger protein basonuclin-2 | |
Rabbit | |
116 kDa | |
100 μL | |
Primary | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS & 2% Sucrose. with 0.09% Sodium Azide | |
BNC2 | |
Synthetic peptides corresponding to Bnc2 (basonuclin 2) The peptide sequence was selected from the C terminal of Bnc2. Peptide sequence GTLRVHYKTVHLREMHKCKVPGCNMMFSSVRSRNRHSQNPNLHKNIPFTS. | |
Affinity Purified | |
RUO | |
54796 | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title