Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BNIP1 Antibody (OTI2B3), DyLight 680, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP270257FR

Catalog No. NBP270257FR


Add to Cart

Description

Description

BNIP1 Monoclonal specifically detects BNIP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

BNIP1
Monoclonal
DyLight 680
PBS with 0.05% Sodium Azide
BNIP1
Recombinant protein fragment corresponding to amino acids 1-199 of human BNIP1 (NP_001196) produced in E.coli.
0.1 mL
Primary
Human, Mouse, Rat
IgG1
Western Blot, Immunohistochemistry
OTI2B3
Western Blot, Immunohistochemistry
BCL2/adenovirus E1B 19 kDa protein-interacting protein 1, BCL2/adenovirus E1B 19kDa interacting protein 1, BCL2/adenovirus E1B 19kD-interacting protein 1, NIP1, SEC20, SEC20L, Transformation-related gene 8 protein, TRG-8, vesicle transport protein SEC20
Mouse
Affinity Purified
Apoptosis
662.0
Store at 4°C in the dark.
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit