Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BNIP1 Mouse anti-Human, Clone: OTI2B3, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526528

Catalog No. PIMA526528


Add to cart

Description

Description

NIP1 (BNIP1) is a member of the BCL2/adenovirus E1B 19 kd-interacting protein (BNIP) family. It interacts with the E1B 19 kDa protein which is responsible for the protection of virally-induced cell death, as well as E1B 19 kDa-like sequences of BCL2, also an apoptotic protector. Alternative splicing of this gene results in four products of unknown function. Transcript variant BNIP1 contains the entire coding region of the gene. This variant contains a fully conserved BH3 domain, which has been associated with pro-apoptotic function.
Specifications

Specifications

BNIP1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q12981
BNIP1; BCL2/adenovirus E1B 19kDa interacting protein 1; NIP1; SEC20; TRG-8; BCL2/adenovirus E1B 19kD interacting protein 1; BCL2/adenovirus E1B 19kD-interacting protein 1
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
662
Immunohistochemistry (Paraffin), Western Blot
OTI2B3
Unconjugated
BNIP1
Liquid
BNIP1
Human recombit protein fragment corresponding to amino acids 1-199 of BNIP1 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit