Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BNIP2 Mouse anti-Human, Alexa Fluor™ 405, Clone: OTI1C12, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270258AF405

Catalog No. N270258A405


Add to cart

Description

Description

BNIP2 Monoclonal antibody specifically detects BNIP2 in Human samples. It is validated for Western Blot.
Specifications

Specifications

BNIP2
OTI1C12
Western Blot
Affinity Purified, Conjugated
BNIP2
Full length recombinant protein of human BNIP2 (NP_004321) produced in HEK293T cells.
Immunogen affinity purified
Apoptosis
Primary
663.0
Western Blot
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
BCL2/adenovirus E1B 19 kDa protein-interacting protein 2, BCL2/adenovirus E1B 19kDa interacting protein 2, BCL2/adenovirus E1B 19kD-interacting protein 2, BNIP-2, NIP2
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit