Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BNIP3 Mouse anti-Human, Alexa Fluor™ 532, Clone: ANa40, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NB1001543AF532

Catalog No. N101543A532


Add to cart

Description

Description

BNIP3 Monoclonal antibody specifically detects BNIP3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

BNIP3
ANa40
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Conjugated, Purified
BNIP3
Recombinant fragment (Residues 1-163 of Human BNIP3).
Protein A or G purified
Apoptosis, Autophagy, Cancer, Hypoxia, Phospho Specific, Tumor Suppressors
Primary
664.0
Human
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
BCL2/adenovirus E1B 19 kDa protein-interacting protein 3, BCL2/adenovirus E1B 19kDa interacting protein 3, BCL2/adenovirus E1B 19kD-interacting protein 3, NIP3
Mouse
IgG2b
0.1 mL
Store at 4°C in the dark.
Monoclonal
BNIP3 (ANa40)
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit