Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BNIPL Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP17950220UL

 View more versions of this product

Catalog No. NBP17950220


Add to cart

Description

Description

BNIPL Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

BNIPL
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
BNIPL
Synthetic peptide directed towards the middle region of human BNIPLThe immunogen for this antibody is BNIPL. Peptide sequence AENYLLVHLSGGTSRAQVPPLSWIRQCYRTLDRRLRKNLRALVVVHATWY.
40 kDa
20ul
Cytokine Research
Primary
149428
Western Blot
Western Blot 1:1000
NP_612122
bcl-2/adenovirus E1B 19 kDa-interacting protein 2-like protein, BCL2/adenovirus E1B 19kD interacting protein like, BNIP-2 similar, BNIPl-1, BNIPL-2, BNIP-S, PP753
Rabbit
IgG
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit