Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BNTX maleate, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

864461-31-4
C31H31NO8
545.588
GKHUUKGUNNYVGB-RAJBGOKPSA-N
7-Benzylidenenaltrexone maleate
124081000
(4R,4aR,6E,7aR,12bR)-6-benzylidene-3-(cyclopropylmethyl)-4a,9-dihydroxy-1,2,4,5,7a,13-hexahydro-4,12-methanobenzofuro[3,2-e]isoquinoline-7-one;(E)-but-2-enedioic acid
C1CC1CN2CCC34C5C(=O)C(=CC6=CC=CC=C6)CC3(C2CC7=C4C(=C(C=C7)O)O5)O.C(=CC(=O)O)C(=O)O

Standard δ1 selective antagonist

Standard selective δ1 opioid receptor antagonist. Also inhibits neurogenic ion transport mediated by a putative novel opioid receptor in porcine ileal mucosa.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 50mg
Formula Weight 545.59
Chemical Name or Material BNTX maleate
Grade >99%

Recommended Storage : Desiccate at -20°C