Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BOB1, Mouse anti-Human, Biotin, Clone: BOB1/2421, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP279885B
Description
BOB1 Monoclonal antibody specifically detects BOB1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
BOB1 | |
BOB1/2421 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Conjugated, Purified | |
POU2AF1 | |
Recombinant fragment (around aa 148-255) of human BOB1 protein (exact sequence is proprietary) | |
Protein A or G purified | |
RUO | |
Primary | |
Human |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Biotin | |
PBS with 0.05% Sodium Azide | |
B-cell-specific coactivator OBF-1, BOB-1, OBF-1, OBF1BOB1, OCAB, OCA-B, OCT-binding factor 1, POU class 2 associating factor 1, POU domain class 2, associating factor 1, POU domain class 2-associating factor 1, POU domain, class 2, associating factor 1 | |
Mouse | |
IgG2b Kappa | |
0.1 mL | |
Store at 4°C in the dark. | |
Monoclonal |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title