Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BOB1, Mouse anti-Human, Clone: BOB1/2421, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP27975020UG
Description
BOB1 Monoclonal specifically detects BOB1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
BOB1 | |
Monoclonal | |
0.2 mg/mL | |
Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/ml, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
Q16633 | |
POU2AF1 | |
Recombinant fragment (around aa 148-255) of human BOB1 protein (exact sequence is proprietary) | |
20 μg | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
BOB1/2421 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
B-cell-specific coactivator OBF-1, BOB-1, OBF-1, OBF1BOB1, OCAB, OCA-B, OCT-binding factor 1, POU class 2 associating factor 1, POU domain class 2, associating factor 1, POU domain class 2-associating factor 1, POU domain, class 2, associating factor 1 | |
Mouse | |
Protein A or G purified | |
RUO | |
5450 | |
Store at 4°C. | |
IgG2b κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title