Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BOB1, Mouse anti-Human, Janelia Fluor 549, Clone: BOB1/2424, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP275753JF549
Description
BOB1 Monoclonal antibody specifically detects BOB1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
BOB1 | |
BOB1/2424 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
50mM Sodium Borate with 0.05% Sodium Azide | |
POU2AF1 | |
Recombinant human BOB1 (POU2AF1) protein fragment (around aa 148-255) (exact sequence is proprietary) | |
Monoclonal | |
Protein A or G purified | |
RUO | |
Human |
Western Blot,Flow Cytometry,Immunohistochemistry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin) | |
Janelia Fluor 549 | |
Purified,Conjugated | |
B-cell-specific coactivator OBF-1, BOB-1, OBF-1, OBF1BOB1, OCAB, OCA-B, OCT-binding factor 1, POU class 2 associating factor 1, POU domain class 2, associating factor 1, POU domain class 2-associating factor 1, POU domain, class 2, associating factor 1 | |
Mouse | |
IgG2b Kappa | |
Primary | |
0.1 mL | |
Store at 4°C in the dark. | |
BOB1 expression in a variety of established B-cell lines, representing different stages of B-cell development, has suggested a constitutive, B-cell-specific expression pattern. LP cells in nodular lymphocyte predominant Hodgkin lymphoma, because they are germinal center-derived, are consistently immuno-positive for BOB1. Conversely, only some cases of classical Hodgkin lymphoma show BOB1 immuno-reactivity within the Hodgkin and Reed-Sternberg cells. Expression of BOB1 has been reported in follicular center cell lymphoma, diffuse large B-cell lymphoma and some cases of acute myeloid leukemia. B-CLL, marginal zone lymphoma, and mantle cell lymphoma may show weak to moderate immunoreactivity. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title