Learn More
Chem-Impex International, Inc.Supplier Diversity Partner Boc-L-alanine, 15761-38-3, MFCD00037225, 25g DFS Item
Manufacturer:  Chem-Impex International, Inc.Supplier Diversity Partner 0132225G
Molecular Formula C8H15NO4, Purity Limit ≥ 99.5% (Chiral HPLC), Molecular Weight 189.20, Melting Point 78 - 85 °C, Synonyms: Boc-L-Ala-OH
Catalog No. 50-493-035