Alfa Aesar™ (Boc-aminooxy)acetic acid, 98+%

Manufacturer: Alfa Aesar™ H5410914

Quantity 25g

 View more versions of this product

Catalog No. AAH5410914

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 42989-85-5
    Molecular Formula C7H13NO5
    Linear Formula (CH3)3COCONHOCH2CO2H
    Formula Weight 191.18
    MDL Number MFCD01632027
    Beilstein 6137646
    Synonym (tert-Butoxycarbonylaminooxy)acetic acid
    Chemical Name or Material (Boc-aminooxy)acetic acid

    Specifications

    Formula Weight 191.18
    Molecular Formula C7H13NO5
    Melting Point ≈115°C
    Linear Formula (CH3)3COCONHOCH2CO2H
    MDL Number MFCD01632027
    Quantity 25g
    Synonym (tert-Butoxycarbonylaminooxy)acetic acid
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 42989-85-5
    Beilstein 6137646
    Chemical Name or Material (Boc-aminooxy)acetic acid
    Assay ≥98%

    Description

    Applications
    (Boc-aminooxy)acetic acid was employed to introduce a hydroxylamine moiety into peptides; reaction with an aldehyde to an oxime. It was used in the preparation of Boc-aminooxy tetra(ethylene glycol).

    Solubility
    Soluble in methanol.

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit