Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boc-Asn Trt -OH, Advanced ChemTech

Catalog No. p-200024073
Click to view available options
Quantity:
1 g
5 g
25 g

Chemical Identifiers

CAS 132388-68-2
Molecular Formula C28H30N2O5
Molecular Weight (g/mol) 474.56
InChI Key PYGOCFDOBSXROC-UHFFFAOYNA-N
IUPAC Name 2-{[(tert-butoxy)carbonyl]amino}-3-[(triphenylmethyl)carbamoyl]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)NC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C(O)=O

Specifications

Color White to Off-White
Quantity 5 g
Grade Biochemical
Chemical Name or Material Boc-Asn Trt -OH
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.