Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BOC-D-Aspartic acid β-benzyl ester, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
51186-58-4
C16H21NO6
323.35
SOHLZANWVLCPHK-GFCCVEGCSA-N
boc-d-asp obzl-oh,boc-d-aspartic acid 4-benzyl ester,r-4-benzyloxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid,2r-4-benzyloxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid,n-alpha-t-boc-d-aspartic acid beta-benzyl ester,boc-d-asparticacid4-benzylester,pubchem11179,boc-d-asp obn-oh,boc-d-aspartic acid b-benzyl ester,boc-d-aspartic acid ?-benzyl ester
6994663
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-phenylmethoxybutanoic acid
CC(C)(C)OC(=O)NC(CC(=O)OCC1=CC=CC=C1)C(=O)O
51186-58-4
C16H21NO6
323.35
SOHLZANWVLCPHK-GFCCVEGCSA-N
boc-d-asp obzl-oh,boc-d-aspartic acid 4-benzyl ester,r-4-benzyloxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid,2r-4-benzyloxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid,n-alpha-t-boc-d-aspartic acid beta-benzyl ester,boc-d-asparticacid4-benzylester,pubchem11179,boc-d-asp obn-oh,boc-d-aspartic acid b-benzyl ester,boc-d-aspartic acid ?-benzyl ester
6994663
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-phenylmethoxybutanoic acid
CC(C)(C)OC(=O)NC(CC(=O)OCC1=CC=CC=C1)C(=O)O
Melting Point 101°C to 105°C
Specific Rotation 19.56
Quantity 5g
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 323.35
Physical Form Powder
Percent Purity 98%
Chemical Name or Material BOC-D-Aspartic acid β-benzyl ester