Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BOC-D-Leucine monohydrate, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
16937-99-8
C11H23NO5
249.307
MFCD00065583
URQQEIOTRWJXBA-DDWIOCJRSA-N
boc-d-leucine monohydrate,r-2-tert-butoxycarbonyl amino-4-methylpentanoic acid hydrate,boc-d-leu-oh.h2o,n-boc-d-leucine monohydrate,boc-d-leu-oh h2o,d-leucine, n-1,1-dimethylethoxy carbonyl-, monohydrate,2r-2-tert-butoxycarbonyl amino-4-methylpentanoic acid hydrate,boc-d-leucine h2o,boc-d-leucine hydrate,n-boc-d-leucinemonohydrate
2724763
(2R)-4-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid;hydrate
CC(C)CC(C(=O)O)NC(=O)OC(C)(C)C.O
16937-99-8
C11H23NO5
249.307
MFCD00065583
URQQEIOTRWJXBA-DDWIOCJRSA-N
boc-d-leucine monohydrate,r-2-tert-butoxycarbonyl amino-4-methylpentanoic acid hydrate,boc-d-leu-oh.h2o,n-boc-d-leucine monohydrate,boc-d-leu-oh h2o,d-leucine, n-1,1-dimethylethoxy carbonyl-, monohydrate,2r-2-tert-butoxycarbonyl amino-4-methylpentanoic acid hydrate,boc-d-leucine h2o,boc-d-leucine hydrate,n-boc-d-leucinemonohydrate
2724763
(2R)-4-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid;hydrate
CC(C)CC(C(=O)O)NC(=O)OC(C)(C)C.O
Assay Percent Range 98%
Linear Formula (CH3)2CHCH2CH(COOH)NHCOOC(CH3)3·H2O
Packaging Glass bottle
Formula Weight 249.31
Quantity 1g
Chemical Name or Material BOC-D-Leucine monohydrate, 98%