Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BOC-D-Methionine, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
25g
Packaging:
Glass bottle
5241-66-7
C10H19NO4S
249.325
MFCD00038256
IMUSLIHRIYOHEV-SSDOTTSWSA-N
boc-d-met-oh,boc-d-methionine,n-boc-d-methionine,n-tert-butoxycarbonyl-d-methionine,n-tert-butyloxycarbonyl-d-methionine,boc-l-met-oh,d-methionine, n-1,1-dimethylethoxy carbonyl,tert-butoxycarbonyl-d-methionine,n-alpha-t-butyloxycarbonyl-d-methionine
111051
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-methylsulfanylbutanoic acid
CC(C)(C)OC(=O)NC(CCSC)C(=O)O
5241-66-7
C10H19NO4S
249.325
MFCD00038256
IMUSLIHRIYOHEV-SSDOTTSWSA-N
boc-d-met-oh,boc-d-methionine,n-boc-d-methionine,n-tert-butoxycarbonyl-d-methionine,n-tert-butyloxycarbonyl-d-methionine,boc-l-met-oh,d-methionine, n-1,1-dimethylethoxy carbonyl,tert-butoxycarbonyl-d-methionine,n-alpha-t-butyloxycarbonyl-d-methionine
111051
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-methylsulfanylbutanoic acid
CC(C)(C)OC(=O)NC(CCSC)C(=O)O
Color White
Specific Rotation Condition +23.00 (20.00°C c=1.3,methanol)
Specific Rotation +21.5° to +24.5° (20°C, 589nm) (c=1, CH3OH)
Packaging Glass bottle
Infrared Spectrum Authentic
Water 0.5% max. (K.F.)
Melting Point 49°C to 55°C
Quantity 25g
Assay Percent Range 98%
Linear Formula CH3SCH2CH2CH(COOH)NHCOOC(CH3)3
Formula Weight 249.32
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material BOC-D-Methionine
Show More Show Less