Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BOC-D-Proline, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
37784-17-1
C10H17NO4
215.249
MFCD00063226
ZQEBQGAAWMOMAI-SSDOTTSWSA-N
n-boc-d-proline,boc-d-proline,boc-d-pro-oh,r-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid,n-tert-butoxycarbonyl-d-proline,2r-1-tert-butoxy carbonyl pyrrolidine-2-carboxylic acid,n-t-boc-d-proline,boc-d-pro,n-alpha-t-boc-d-proline,1-tert-butoxycarbonyl-d-proline
688022
(2R)-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid
CC(C)(C)OC(=O)N1CCCC1C(=O)O
37784-17-1
C10H17NO4
215.249
MFCD00063226
ZQEBQGAAWMOMAI-SSDOTTSWSA-N
n-boc-d-proline,boc-d-proline,boc-d-pro-oh,r-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid,n-tert-butoxycarbonyl-d-proline,2r-1-tert-butoxy carbonyl pyrrolidine-2-carboxylic acid,n-t-boc-d-proline,boc-d-pro,n-alpha-t-boc-d-proline,1-tert-butoxycarbonyl-d-proline
688022
(2R)-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid
CC(C)(C)OC(=O)N1CCCC1C(=O)O
Assay Percent Range 98%
Infrared Spectrum Authentic
Packaging Glass bottle
Formula Weight 215.25
Specific Rotation +59° to +62° (20°C, 589nm) (c=1, CH3COOH)
Physical Form Crystalline Powder or Crystals
Percent Purity ≥97.5%
Color White
Melting Point >132°C
Quantity 1g
Chemical Name or Material BOC-D-Proline, 98%
Show More Show Less