Learn More
Edwards Rotary Vane Vacuum Pumps for Centrifugal Concentrators and Rotary Evaporators: RV12
Deliver high or low throughputs with ultimate high vacuum, which combined with ultra-low noise levels
Manufacturer:  Edwards R44028601
Catalog No. 11-283-101
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Dual-mode
  • Adjustable 3-position gas ballast
  • Integral air bleed
  • NW25 inlet/outlet connect
  • Wide voltage motors
  • IEC line cord
  • Patented inlet valve and mode switch
  • Oil tight printed gasket
Height includes attached lifting bracket.
Certifications
UL listed, CSA standards
Specifications
Specifications
6 x 10-2mbar
3 Position gas ballast, NW25 inlet/outlet connection, IEC line cord, 3/4hp motor
UL, CSA
238dBA
58 lb.
158°F
50°C
Centrifugal concentrators/rotary evaporator
8.9 in.
19.3 in.
6.2 in.
0.75hp
1500 to 1800
RV12
-30 to 70°C
26.3kg
70°C
283L/min.
50/60Hz
22.5cm
49cm
IP44
Documents
Accessories